Orders LinkContactUsLinkComponent
Celtellers, Thoma-New
Counting chamber with double net ruling, clamps and bright lines, Thoma new, 0,1 mm
  HIRS8110206
 :   HIRSCHMANN
Celtellers, Thoma-New
  HIRS8110206
Celtellers, Thoma-New
Celtellers, Thoma-New
Counting chamber with double net ruling, clamps and bright lines, Thoma new, 0,1 mm
178,00 €
1 * 1 ST
Thoma new counting chambers with double net ruling.
 : IVD 98/79 EC
 : Include two cover glasses for haemacytometer.
  • Kamerdiepte:
    0,1 mm
  • Type:
    Thoma new
  • Omschrijving:
    Counting chamber with double net ruling, clamps and bright lines