Orders LinkContactUsLinkComponent
Celtellers, Thoma
Celtellers, Thoma
Counting chamber, double ruling, bright-lined, with clips, Thoma, 0,10 mm
Catalogus # 612-5731
Leverancier:   HIRSCHMANN
Celtellers, Thoma
Catalogus # 612-5731
Leverancier:  HIRSCHMANN
Celtellers, Thoma
Celtellers, Thoma
Counting chamber, double ruling, bright-lined, with clips, Thoma, 0,10 mm
Price
178,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Cell counting chambers including two cover glasses for haemacytometers.
Specificaties
  • Kamerdiepte:
    0,10 mm
  • Omschrijving:
    Counting chamber, double ruling, bright-lined, with clips, Thoma