Celtellers, Thoma
Counting chamber, double ruling, bright-lined, with clips, Thoma, 0,10 mm
Catalogus # 612-5731
Leverancier: HIRSCHMANN
Ga naar:
Product Details & Documents
Cell counting chambers including two cover glasses for haemacytometers.
Specificaties
- Kamerdiepte:0,10 mm
- Omschrijving:Counting chamber, double ruling, bright-lined, with clips, Thoma
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.