Celtellers, Thoma-New
Celteller, dubbele nettotelling, met klemmen, Thoma new, 0,1 mm
Catalogus # HIRS8110106
Leverancier: HIRSCHMANN
Ga naar:
Product Details & Documents
Thoma new counting chambers with double net ruling.
: IVD 98/79 EC
: Include two cover glasses for haemacytometer.
Specificaties
- Kamerdiepte:0,1 mm
- Type:Thoma new
- Omschrijving:Celteller, dubbele nettotelling, met klemmen