Orders LinkContactUsLinkComponent
Celtellers, Fuchs-Rosenthal
Celtellers, Fuchs-Rosenthal
Cell counter, bright-lined, 0,2 mm
Catalog # HIRS8100210
Supplier:   HIRSCHMANN
Celtellers, Fuchs-Rosenthal
Catalog # HIRS8100210
Supplier:  HIRSCHMANN
Celtellers, Fuchs-Rosenthal
Celtellers, Fuchs-Rosenthal
Cell counter, bright-lined, 0,2 mm
Price
175,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Telkamers met dubbele nettotelling.
Ordering information: Includes two cover glasses.
Specifications
  • Kamerdiepte:
    0,2 mm
  • Omschrijving:
    Cell counter, bright-lined