Celtellers, Fuchs-Rosenthal
Cell counter, bright-lined, 0,2 mm
Catalog # HIRS8100210
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
Telkamers met dubbele nettotelling.
Ordering information: Includes two cover glasses.
Specifications
- Kamerdiepte:0,2 mm
- Omschrijving:Cell counter, bright-lined
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.