Orders LinkContactUsLinkComponent
Celtellers, Thoma-New
Celtellers, Thoma-New
Counting chamber with double net ruling and bright lines, Thoma new, 0,1 mm
Catalog # HIRS8100206
Supplier:   HIRSCHMANN
Celtellers, Thoma-New
Catalog # HIRS8100206
Supplier:  HIRSCHMANN
Celtellers, Thoma-New
Celtellers, Thoma-New
Counting chamber with double net ruling and bright lines, Thoma new, 0,1 mm
Price
159,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Thoma new counting chambers with double net ruling.
Certifications: IVD 98/79 EC
Ordering information: Include two cover glasses for haemacytometer.
Specifications
  • Kamerdiepte:
    0,1 mm
  • Type:
    Thoma new
  • Omschrijving:
    Counting chamber with double net ruling and bright lines