Celtellers, Thoma-New
Counting chamber with double net ruling and bright lines, Thoma new, 0,1 mm
Catalog # HIRS8100206
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
Thoma new counting chambers with double net ruling.
Certifications: IVD 98/79 EC
Ordering information: Include two cover glasses for haemacytometer.
Specifications
- Kamerdiepte:0,1 mm
- Type:Thoma new
- Omschrijving:Counting chamber with double net ruling and bright lines
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.