Celtellers, Thoma
Counting chamber, double ruling, bright-lined, Thoma, 0,10 mm
Catalogus # 612-5726
Leverancier: HIRSCHMANN
Ga naar:
Product Details & Documents
Cell counting chambers including two cover glasses for haemacytometers.
Specificaties
- Kamerdiepte:0,10 mm
- Omschrijving:Counting chamber, double ruling, bright-lined, Thoma
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.