Orders LinkContactUsLinkComponent
Celtellers, Thoma-New
Celtellers, Thoma-New
Counting chamber with double net ruling, Thoma new, 0,1 mm
Catalogus # HIRS8100106
Leverancier:   HIRSCHMANN
Celtellers, Thoma-New
Catalogus # HIRS8100106
Leverancier:  HIRSCHMANN
Celtellers, Thoma-New
Celtellers, Thoma-New
Counting chamber with double net ruling, Thoma new, 0,1 mm
Price
66,90 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Thoma new counting chambers with double net ruling.
 : IVD 98/79 EC
 : Include two cover glasses for haemacytometer.
Specificaties
  • Kamerdiepte:
    0,1 mm
  • Type:
    Thoma new
  • Omschrijving:
    Counting chamber with double net ruling