Orders LinkContactUsLinkComponent
Inlaat liners
Inlaat liners
Capillary inlet trap assembly
Catalogus # HEWLG1544-80550
Leverancier:   Agilent
Inlaat liners
Catalogus # HEWLG1544-80550
Leverancier:  Agilent
Inlaat liners
Inlaat liners
Capillary inlet trap assembly
Price
365,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
  • Consistent high level of inertness
Specificaties
  • Omschrijving:
    Capillary inlet trap assembly