Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Deactivated/silinised vials

Deactivated/silinised vials

Vials, certified, 0,25 ml, 12×32 mm, Neen, Crimp/snap
Catalogus # HEWL9301-0978
Leverancier:   Agilent
Deactivated/silinised vials
Catalogus # HEWL9301-0978
Leverancier:  Agilent
Deactivated/silinised vials
Deactivated/silinised vials

Vials, certified, 0,25 ml, 12×32 mm, Neen, Crimp/snap

Price
355,00 €
Log in voor beschikbaarheid
1 * 1.000 ST
Product Details & Documents

These vials made of PP are used for pH sensitive samples, sodium or heavy metals analysis.

  • Translucent
  • Good chemical resistance
  • Can be used with crimp or snap caps

Vials with glass insert are used for economical and effective microsampling. The PP body has a glass flanged insert molded to the inside, so the sample comes in contact only with the 100 µl glass insert and the septum.

Specificaties
  • Inclusief dop:
    Neen
  • Ø×h:
    12×32 mm
  • Capaciteit:
    0,25 ml
  • Omschrijving:
    Vials, certified
  • Soort dop:
    Crimp/snap