Tools
Hex key, 1,5 mm, 10 cm long, straight handle
Catalogus # HEWL8710-2393
Leverancier: Agilent
Ga naar:
Product Details & Documents
Specificaties
- Omschrijving:Hex key, 1,5 mm, 10 cm long, straight handle