Inlaat liners
Straight, direct inlet liner, non-deactivated (for gas samples headspace or purge & trap), 140 μl, 1,5 mm
Catalogus # HEWL5183-4709
Leverancier: Agilent
Ga naar:
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
Specificaties
- Ø inw.:1,5 mm
- Omschrijving:Straight, direct inlet liner, non-deactivated (for gas samples headspace or purge & trap), 140 μl
- Seq:48