Inlet septa
Merlin microseal kit, original low pressure system (includes nut and septum)
HEWL5181-8816
: Agilent
These septa are available for a variety of applications and have different upper temperature limits. The lower temperature septa are usually softer, seal better and can withstand more punctures (injections) than their high temperature counterparts.
- Made up low bleed silicone rubber formulations
- Efficient by using sharp syringe needles
- Septa should be replaced to avoid leaks and sample loss
- To be used with recommended temperature range
- Omschrijving:Merlin microseal kit, original low pressure system (includes nut and septum)
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.