Inlet septa
Microseal septum, stainless steel, rubber (30 psi)
Catalogus # HEWL5181-8815
Leverancier: Agilent
Ga naar:
Product Details & Documents
These septa are available for a variety of applications and have different upper temperature limits. The lower temperature septa are usually softer, seal better and can withstand more punctures (injections) than their high temperature counterparts.
- Made up low bleed silicone rubber formulations
- Efficient by using sharp syringe needles
- Septa should be replaced to avoid leaks and sample loss
- To be used with recommended temperature range
Specificaties
- Omschrijving:Microseal septum, stainless steel, rubber (30 psi)