Inlet septa
Septa, general purpose, grey, 5 mm
Catalog # HEWL5181-1260
Supplier: Agilent
Jump to:
Product Details & Documents
These septa are available for a variety of applications and have different upper temperature limits. The lower temperature septa are usually softer, seal better and can withstand more punctures (injections) than their high temperature counterparts.
- Made up low bleed silicone rubber formulations
- Efficient by using sharp syringe needles
- Septa should be replaced to avoid leaks and sample loss
- To be used with recommended temperature range
Specifications
- Ø:5 mm
- Omschrijving:Septa, general purpose, grey