Dissecting forceps
Dissecting forceps, stainless steel, tip width 0,05 mm, tip thickness 0,01 mm, Straight, 110 mm
Catalogus # HARV60-3854
Leverancier: HARVARD APPARATUS
Ga naar:
Product Details & Documents
Micro dissecting forceps with tower tip. Made from stainless steel or Inox steel.
Specificaties
- Type:Straight
- Omschrijving:Dissecting forceps, stainless steel, tip width 0,05 mm, tip thickness 0,01 mm
- Lengte:110 mm