Dissecting forceps
Dissecting forceps, Inox steel, tip width 0,10 mm, tip thickness 0,06 mm, Straight, 110 mm
HARV52-2060
Micro dissecting forceps with tower tip. Made from stainless steel or Inox steel.
- Type:Straight
- Omschrijving:Dissecting forceps, Inox steel, tip width 0,10 mm, tip thickness 0,06 mm
- Lengte:110 mm