Orders LinkContactUsLinkComponent
Lepels
Lepels
205 mm, 70×55 mm
Catalog # HAMMHWN144-20
Supplier:   HAMMACHER KARL
Lepels
Catalog # HAMMHWN144-20
Supplier:  HAMMACHER KARL
Lepels
Lepels
205 mm, 70×55 mm
Price
52,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Uit antimagnetisch roestvrij staal 18/10.
Specifications
  • Lengte:
    205 mm
  • Lepelmaat:
    70×55 mm