Serrefines
Serrefine, Straight, 35 mm
Catalogus # 221-0051
Leverancier: HAMMACHER KARL
Ga naar:
Product Details & Documents
These small forceps for clamping a blood vessel are made from stainless steel.
Specificaties
- Type:Straight
- Omschrijving:Serrefine
- Lengte:35 mm
- Seq:03
- Cat. No.:221-0051