Phosphate standard solutions
Phosphate standard solution, 946 ml (NIST), 3 mg/l as PO43-, 946 mL
HACH2059716
: Hach
Standard solution for accuracy checks in phosphate (phosphorus) determinations.
- Concentratie:3 mg/l as PO<sub>4</sub><sup>3-</sup>
- VPE:946 mL
- Omschrijving:Phosphate standard solution, 946 ml (NIST)



