Orders LinkContactUsLinkComponent
Redox standards
Redox standards
Standard redox solution, 650 mV at 25 °C, 500 mL
Catalogus # REGERS650
Leverancier:   REAGECON
Redox standards
Catalogus # REGERS650
Leverancier:  REAGECON
CAS-nummer:  
Redox standards
Standard redox solution, 650 mV at 25 °C, 500 mL
CAS-nummer:  
Price
108,00 €
Log in voor beschikbaarheid
1 * 500 mL
Product Details & Documents
The potentials quoted are of platinum electrode with reference to Ag/AgCl (3 M KCl).
Specificaties
Specificaties
  • VPE:
    500 mL
  • Omschrijving:
    Standard redox solution, 650 mV at 25 °C
Specificaties