Orders LinkContactUsLinkComponent
Redox standards
Redox standards
Standard redox solution, 124 mV at 25 °C, 500 mL
Catalog # REGERS124
Supplier:   REAGECON
Redox standards
Catalog # REGERS124
Supplier:  REAGECON
CAS Number:  
Redox standards
Standard redox solution, 124 mV at 25 °C, 500 mL
CAS Number:  
Price
108,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
The potentials quoted are of platinum electrode with reference to Ag/AgCl (3 M KCl).
Documents
Specifications
Specifications
  • VPE:
    500 mL
  • Omschrijving:
    Standard redox solution, 124 mV at 25 °C
Specifications