Orders LinkContactUsLinkComponent
COD standards
COD standard solution, 600 mg/l, 500 mL
  REGECOD600
 :   REAGECON
COD standards
  REGECOD600
 :  REAGECON
 :  
COD standards
COD standard solution, 600 mg/l, 500 mL
 :  
198,00 €
1 * 500 mL
Ideal for use as control standards to verify that correct analysis for COD has taken place.
  • Concentratie:
    600 mg/l
  • VPE:
    500 mL
  • Omschrijving:
    COD standard solution