Biology prepared microscope slides, school set D
Frog embryology (Rana spec.) section trough the blastula stage showing the blastocoel
Catalogus # LIED791F
Leverancier: LIEDER JOHANNES
Ga naar:
Product Details & Documents
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
- Microtome sections individually crafted under rigorous scientific control
- Each slide carefully selected on the basis of instructional value
- Multicolour staining for distinct variation
The sets contain slides of typical micro-organisms, of cell division and of embryonic developments, as well as of tissues and organs of plants, animals and man.
Levering: Supplied on high quality ground edge slides.
Specificaties
- Omschrijving:Frog embryology (Rana spec.) section trough the blastula stage showing the blastocoel
- Seq:429
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.