Orders LinkContactUsLinkComponent
Biology prepared microscope slides, school set D
Sea-urchin development (Psammechinus miliaris) composite slide with morula, blastula and gastrula stages
  LIED790E
Biology prepared microscope slides, school set D
  LIED790E
Biology prepared microscope slides, school set D
Biology prepared microscope slides, school set D
Sea-urchin development (Psammechinus miliaris) composite slide with morula, blastula and gastrula stages
10,20 €
1 * 1 ST
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
  • Microtome sections individually crafted under rigorous scientific control
  • Each slide carefully selected on the basis of instructional value
  • Multicolour staining for distinct variation
The sets contain slides of typical micro-organisms, of cell division and of embryonic developments, as well as of tissues and organs of plants, animals and man.
 : Supplied on high quality ground edge slides.
  • Omschrijving:
    Sea-urchin development (Psammechinus miliaris) composite slide with morula, blastula and gastrula stages
  • Seq:
    428