Biology prepared microscope slides, school set D
Sea-urchin development (Psammechinus miliaris) composite slide with morula, blastula and gastrula stages
LIED790E
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
- Microtome sections individually crafted under rigorous scientific control
- Each slide carefully selected on the basis of instructional value
- Multicolour staining for distinct variation
The sets contain slides of typical micro-organisms, of cell division and of embryonic developments, as well as of tissues and organs of plants, animals and man.
: Supplied on high quality ground edge slides.
- Omschrijving:Sea-urchin development (Psammechinus miliaris) composite slide with morula, blastula and gastrula stages
- Seq:428