Biology prepared microscope slides, school set D
Sea-urchin development (Psammechinus miliaris) composite slide with two cell-, four cell- and eight cell-stages
LIED789E
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
- Microtome sections individually crafted under rigorous scientific control
- Each slide carefully selected on the basis of instructional value
- Multicolour staining for distinct variation
The sets contain slides of typical micro-organisms, of cell division and of embryonic developments, as well as of tissues and organs of plants, animals and man.
: Supplied on high quality ground edge slides.
- Omschrijving:Sea-urchin development (Psammechinus miliaris) composite slide with two cell-, four cell- and eight cell-stages
- Seq:427
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.