Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Biology prepared microscope slides, school set D
Biology prepared microscope slides, school set D
DNA and RNA fixed and stained with methyl green and pyronine staining
Catalogus # LIED776G
Leverancier:   LIEDER JOHANNES
Biology prepared microscope slides, school set D
Catalogus # LIED776G
Leverancier:  LIEDER JOHANNES
Biology prepared microscope slides, school set D
Biology prepared microscope slides, school set D
DNA and RNA fixed and stained with methyl green and pyronine staining
Price
16,80 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
  • Microtome sections individually crafted under rigorous scientific control
  • Each slide carefully selected on the basis of instructional value
  • Multicolour staining for distinct variation
The sets contain slides of typical micro-organisms, of cell division and of embryonic developments, as well as of tissues and organs of plants, animals and man.
Levering: Supplied on high quality ground edge slides.
Specificaties
  • Omschrijving:
    DNA and RNA fixed and stained with methyl green and pyronine staining
  • Seq:
    417
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.