Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Biology prepared microscope slides, school set D
DNA in cell nuclei, demonstrated by Feulgen staining technique
Catalog # LIED775G
Supplier:   LIEDER JOHANNES
Biology prepared microscope slides, school set D
Catalog # LIED775G
Supplier:  LIEDER JOHANNES
Biology prepared microscope slides, school set D
Biology prepared microscope slides, school set D
DNA in cell nuclei, demonstrated by Feulgen staining technique
Price
16,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
  • Microtome sections individually crafted under rigorous scientific control
  • Each slide carefully selected on the basis of instructional value
  • Multicolour staining for distinct variation
The sets contain slides of typical micro-organisms, of cell division and of embryonic developments, as well as of tissues and organs of plants, animals and man.
 : Supplied on high quality ground edge slides.
Specifications
  • Omschrijving:
    DNA in cell nuclei, demonstrated by Feulgen staining technique
  • Seq:
    416