Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Biology prepared microscope slides, school set D
Golgi apparatus transverse section through spinal ganglion with special staining technique
Catalogus # LIED771G
Leverancier:   LIEDER JOHANNES
Biology prepared microscope slides, school set D
Catalogus # LIED771G
Leverancier:  LIEDER JOHANNES
Biology prepared microscope slides, school set D
Biology prepared microscope slides, school set D
Golgi apparatus transverse section through spinal ganglion with special staining technique
Price
16,80 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
  • Microtome sections individually crafted under rigorous scientific control
  • Each slide carefully selected on the basis of instructional value
  • Multicolour staining for distinct variation
The sets contain slides of typical micro-organisms, of cell division and of embryonic developments, as well as of tissues and organs of plants, animals and man.
 : Supplied on high quality ground edge slides.
Specificaties
  • Omschrijving:
    Golgi apparatus transverse section through spinal ganglion with special staining technique
  • Seq:
    414
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.