Biology prepared microscope slides, school set D
Mitochondria in thin section through liver or kidney with special staining technique
LIED770F
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
- Microtome sections individually crafted under rigorous scientific control
- Each slide carefully selected on the basis of instructional value
- Multicolour staining for distinct variation
The sets contain slides of typical micro-organisms, of cell division and of embryonic developments, as well as of tissues and organs of plants, animals and man.
: Supplied on high quality ground edge slides.
- Omschrijving:Mitochondria in thin section through liver or kidney with special staining technique
- Seq:413