Biology prepared microscope slides, school set B
Root tip and hair
Catalogus # LIED646D
Leverancier: LIEDER JOHANNES
Biology prepared microscope slides, school set B
Catalogus # LIED646D
Leverancier: LIEDER JOHANNES
Ga naar:
Product Details & Documents
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
- Microtome sections individually crafted under rigorous scientific control
- Each slide carefully selected on the basis of instructional value
- Multicolour staining for distinct variation
The set contains slides of typical micro-organisms, cell division and embryonic developments, as well as of tissues and organs of plants, animals and man.
Delivery information: Supplied on high quality ground edge slides (26×76 mm).
Specificaties
- Omschrijving:Root tip and hair
- Seq:230
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.