Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Biology prepared microscope slides, school set B
Biology prepared microscope slides, school set B
Dahlia tuber with inuline crystals
Catalogus # LIED637D
Leverancier:   LIEDER JOHANNES
Biology prepared microscope slides, school set B
Catalogus # LIED637D
Leverancier:  LIEDER JOHANNES
Biology prepared microscope slides, school set B
Biology prepared microscope slides, school set B
Dahlia tuber with inuline crystals
Price
8,90 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
  • Microtome sections individually crafted under rigorous scientific control
  • Each slide carefully selected on the basis of instructional value
  • Multicolour staining for distinct variation
The set contains slides of typical micro-organisms, cell division and embryonic developments, as well as of tissues and organs of plants, animals and man.
Levering: Supplied on high quality ground edge slides (26×76 mm).
Specificaties
  • Omschrijving:
    Dahlia tuber with inuline crystals
  • Seq:
    224
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.