Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Biology prepared microscope slides, school set B
Biology prepared microscope slides, school set B
Lung of cat alveoli and bronchial tubes
Catalogus # LIED627C
Leverancier:   LIEDER JOHANNES
Biology prepared microscope slides, school set B
Catalogus # LIED627C
Leverancier:  LIEDER JOHANNES
Biology prepared microscope slides, school set B
Biology prepared microscope slides, school set B
Lung of cat alveoli and bronchial tubes
Price
8,60 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
  • Microtome sections individually crafted under rigorous scientific control
  • Each slide carefully selected on the basis of instructional value
  • Multicolour staining for distinct variation
The set contains slides of typical micro-organisms, cell division and embryonic developments, as well as of tissues and organs of plants, animals and man.
Delivery information: Supplied on high quality ground edge slides (26×76 mm).
Specificaties
  • Omschrijving:
    Lung of cat alveoli and bronchial tubes
  • Seq:
    218
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.