PH-combinatie-elektroden
Sensolyt® PtA ORP-meetketting met enkele ketting**, Sensolyt® PtA (ORP), voor installatie in de SensoLyt®-fitting, ±2000 mV, , Sterk verontreinigd afvalwater
Catalogus # 708-0037
Leverancier: WTW ONLINE
Ga naar:
Product Details & Documents
The SensoLyt® measuring chains with arming are easy to install and boast flexible measuring and application areas. Eletrodes without arming can be connected directly to a pH monitor of the EcoLine® or QuadroLine® series. Depending on the application, they can be installed into a flow vessel.
- Ideal for measurements in water and drinking water
- Also for high pressure and temperature conditions
- Easy to maintain by easy measuring chain replacement
Specificaties
- Voor:Sterk verontreinigd afvalwater
- Type:Sensolyt® PtA (ORP), voor installatie in de SensoLyt®-fitting
- Omschrijving:Sensolyt® PtA ORP-meetketting met enkele ketting**
- Seq:38
- pH:±2000 mV
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.