PH-combinatie-elektroden
Sensolyt® SE*, Sensolyt® SE, zonder armering, voor inbouw in doorstroomvaten, 2 - 12, , Sterk verontreinigd afvalwater
Catalogus # 662-0280
Leverancier: WTW ONLINE
Ga naar:
Product Details & Documents
The SensoLyt® measuring chains with arming are easy to install and boast flexible measuring and application areas. Eletrodes without arming can be connected directly to a pH monitor of the EcoLine® or QuadroLine® series. Depending on the application, they can be installed into a flow vessel.
- Ideal for measurements in water and drinking water
- Also for high pressure and temperature conditions
- Easy to maintain by easy measuring chain replacement
Specificaties
- Voor:Sterk verontreinigd afvalwater
- Type:Sensolyt® SE, zonder armering, voor inbouw in doorstroomvaten
- Omschrijving:Sensolyt® SE*
- Seq:31
- pH:2 - 12