Trypsinizing flasks, WHEATON®
75 ml, Schroefdop voor bovenzijde 33-430, Schroefdop 20-415
Catalog # WHEA355803
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Borosilicate glass, clear.
- With pour out and caps
- Fluted design improves mixing
- Deep skirted screw caps
These flasks feature deep-fluted sides to promote mixing. A dam below the sidearm retains untrypsinized tissue when decanting. A deep skirted screw cap on the sidearm prevents contamination of flask contents. The sidearm allows easier decanting of contents while retaining large particles behind the dam. Sidearm caps are 20-415 size. Caps have white styrene-butadiene rubber liners.
Specifications
- Capaciteit:75 ml
- Omschrijving:Schroefdop voor bovenzijde 33-430
- Hals:Schroefdop 20-415