Orders LinkContactUsLinkComponent
Conductivity Standards
Conductivity calibration solution 100 µS/cm, shelf life 2 years, NIST traceable, accuracy ±3%, 300 ml
  662-0292
New Product
 :   WTW
Conductivity Standards
  662-0292
 :  WTW
New Product
 :  
Conductivity Standards
Conductivity Standards
Conductivity calibration solution 100 µS/cm, shelf life 2 years, NIST traceable, accuracy ±3%, 300 ml
 :  
1 * 250 mL
  • VPE:
    300 ml
  • Omschrijving:
    Conductivity calibration solution 100 µS/cm, shelf life 2 years, NIST traceable, accuracy ±3%