EasyPlus™ Titration Electrodes
EG11-BNC sensor pH aqueous, ±2000 mV/0 - 14 pH, Easy Pro, Easy EPM, Easy pH titrators
Catalogus # METT30043103
New Product
Leverancier: Mettler - Toledo
Ga naar:
Product Details & Documents
Electrode for use with Mettler Toledo® range of EasyPlus™ titrators only.
Electrode only.
Documents
How-to Refill an Electrode
Video 's
Specificaties
- Voor:Easy Pro, Easy EPM, Easy pH titrators
- Omschrijving:EG11-BNC sensor pH aqueous
- Seq:1
- Meetbereik:±2000 mV/0 - 14 pH