EasyPlus™ Titration Electrodes
EG13-BNC sensor pH non-aqueous, 0 - 2000 mV, Easy Pro, Easy KFV, Easy Ox titrators
Catalogus # METT30043104
New Product
Leverancier: Mettler - Toledo
Ga naar:
Product Details & Documents
Electrode for use with Mettler Toledo® range of EasyPlus™ titrators only.
Electrode only.
Documents
How-to Refill an Electrode
Video 's
Specificaties
- Voor:Easy Pro, Easy KFV, Easy Ox titrators
- Omschrijving:EG13-BNC sensor pH non-aqueous
- Seq:5
- Meetbereik:0 - 2000 mV