Orders LinkContactUsLinkComponent
Cellroll Set
Cellroll Set
CELLROLL Set, 4 roller bottles for CB 170, CB 170-L, CB 170-O₂, CB 170-O₂L, CBF 170, CBF 170-O₂
Catalogus # BIND8012-1885
New Product
Leverancier:   Binder
Cellroll Set
Catalogus # BIND8012-1885
Leverancier:  Binder
New Product
Cellroll Set
Cellroll Set
CELLROLL Set, 4 roller bottles for CB 170, CB 170-L, CB 170-O₂, CB 170-O₂L, CBF 170, CBF 170-O₂
Price
9 490,00 €
Log in voor beschikbaarheid
Product Details & Documents
The Integra Cellroll is a compact and reliable rotary bottle system for cultivating adherent cells in roller bottles. This modular, expandable roller bottle system is suitable for either 4 or 6 roller bottles.
  • Infinitely variable speed range from 0.1 to 2 rpm, the system enables gentle and uniform rotation of the cell culture bottles Ideal for large-volume cell cultures in research and production Continuous rotation ensures an even distribution of cells and culture medium, which promotes optimal growth conditions and high yields Specially designed for use in CO₂ incubators - with temperature-resistant materials and a compact, stackable design Fine adjustment of rotation direction, rotation speed: Fine adjustment of pause times and interval cycles: With soft start and stop phase:
Specificaties
  • Omschrijving:
    CELLROLL Set, 4 roller bottles for CB 170, CB 170-L, CB 170-O₂, CB 170-O₂L, CBF 170, CBF 170-O₂
  • Seq:
    1