Combination Blocks
MCT-W21-COMBO block, Ø 7,9/11,1/11,5 mm, 0,5/1,5/2,0 ml microcentrifuge tubes, 6/10/5, 27,6/39,1/38,1 mm
Catalogus # VELPA00000513
New Product
Irregular Voltage
Leverancier: VELP SCIENTIFIC
Combination Blocks
Catalogus # VELPA00000513
Leverancier: VELP SCIENTIFIC
New Product
Irregular Voltage
Ga naar:
Product Details & Documents
Specificaties
- Aant. gaten:6/10/5
- Omschrijving:MCT-W21-COMBO block
- Seq:3
- Diepte:27,6/39,1/38,1 mm
- Afmeting opening:Ø 7,9/11,1/11,5 mm
- Voor:0,5/1,5/2,0 ml microcentrifuge tubes
- Cat. No.:VELPA00000513