Orders LinkContactUsLinkComponent
Handheld Digital Microscopes, Universal
Handheld Digital Microscopes, Universal
Handmicroscoop met verwisselbare doppen + zonder polarisator, Edge, UNIVERSAL, AM4815T, 20 - 220×, 8 LED's, 1,3 MP, EDOF/EDR
Catalogus # IDCPAM4815T
New Product
Irregular Voltage
Leverancier:   Dino-Lite / IDCP B.V.
Handheld Digital Microscopes, Universal
Catalogus # IDCPAM4815T
Leverancier:  Dino-Lite / IDCP B.V.
New Product
Irregular Voltage
Handheld Digital Microscopes, Universal
Handheld Digital Microscopes, Universal
Handmicroscoop met verwisselbare doppen + zonder polarisator, Edge, UNIVERSAL, AM4815T, 20 - 220×, 8 LED's, 1,3 MP, EDOF/EDR
Price
642,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
The Universal series provides a broad range of products with high image quality to satisfy the most demanding user. Magnification range: 10 to 220×.
  • High speed USB 2.0 connection
  • Metal or composite housing
  • Edge series models with high quality optics provide a very sharp, bright and natural colour image with very low aberration and vignetting
  • Wireless streaming to a tablet, Smart phone, or computer is possible in combination with WF-10 unit (to be ordered separately)
  • Integrated polariser greatly reduces glare on reflective surfaces (some models only)
Models with Automatic Magnification Reading (AMR): Allows the included software to read the magnification setting from the microscope device, simplifying the measuring process.
Models with Extended Depth of Field (EDOF): Automatically capture and combine multiple images at different focus levels to produce a final image with greater depth of field.
Models with Extended Dynamic Range (EDR): Automatically capture and combine multiple images at different exposure levels to create an image with greater dynamic range.
Models with Flexible LED Control (FLC): Additional LED controls within the included software for adjusting LED brightness and on/off toggle by quadrant.
Delivery information: Supplied with protecting pouch, software CD (DinoCapture 2.0 (Windows), DinoXcope (Mac OS)) and calibration target.
Services: Installatie, IQOQ, onderhoud of reparatie nodig? Mail naar [email protected]
Specificaties
  • Vergroting:
    20 - 220×
  • Model:
    UNIVERSAL, AM4815T
  • Resolutie:
    1,3 MP
  • Omschrijving:
    Handmicroscoop met verwisselbare doppen + zonder polarisator, Edge
  • Seq:
    18
  • Extra functies:
    EDOF/EDR
  • Verlichting:
    8 LED's
  • Cat. No.:
    IDCPAM4815T
Accessories
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Extension pole RK-06-PL, to double the height of the RK-06A / RK-06-AE stand (required when working with Long Working Distance and Extra Long Working Distance microscopes)
    Extension pole RK-06-PL, to double the height of the RK-06A / RK-06-AE stand (required when working with Long Working Distance and Extra Long Working Distance microscopes)
    IDCPRK-06-PL
    Log in voor beschikbaarheid
    1 * 1 ST - 28,60 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Voetpedaal, stelt gebruikers in staat om een foto te maken met hun voet
    Voetpedaal, stelt gebruikers in staat om een foto te maken met hun voet
    630-1821
    Log in voor beschikbaarheid
    1 * 1 ST - 63,30 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Sturdy high-end stand for Dino-Lite microscopes, RK-10A, incl. HD-P1 holder and RK-10-EX horizontal arm
    Sturdy high-end stand for Dino-Lite microscopes, RK-10A, incl. HD-P1 holder and RK-10-EX horizontal arm
    630-2517
    Log in voor beschikbaarheid
    1 * 1 ST - 340,00 €
Productinformatie
  • Catalogus #
    b×d×h
    Beschikbaarheid
    Price
  • Tafelstatief voor handheld Dino-Lite microscopen, RK-06-A, incl. verlengarm RK-10-EX
    Tafelstatief voor handheld Dino-Lite microscopen, RK-06-A, incl. verlengarm RK-10-EX
    IDCPRK-06A
    150×220×270 mm
    Log in voor beschikbaarheid
    1 * 1 ST - 252,00 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Wifi streamer met gebruiksaanwijzing, stroomadapter en 1880 Ah Li-Ion accu
    Wifi streamer met gebruiksaanwijzing, stroomadapter en 1880 Ah Li-Ion accu
    630-2923
    Log in voor beschikbaarheid
    1 * 1 ST - 329,00 €
Productinformatie
  • Catalogus #
    b×d×h
    Beschikbaarheid
    Price
  • Tabletop stand for handheld Dino-Lite microscopes, RK-06-AE, with ESD-safe coating, incl. extension arm RK-10-EX
    Tabletop stand for handheld Dino-Lite microscopes, RK-06-AE, with ESD-safe coating, incl. extension arm RK-10-EX
    IDCPRK-06AE
    150×220×270 mm
    Log in voor beschikbaarheid
    1 * 1 ST - 249,00 €