Phenolphthalein indicator
Bottle
Catalogus # APOSOR1014628-25G
New Product
Leverancier: Apollo Scientific
Phenolphthalein indicator
Catalogus # APOSOR1014628-25G
Leverancier: Apollo Scientific
New Product
CAS-nummer: 77-09-8
Ga naar:
Product Details & Documents
Usage: Organic building block/reagent.
Synoniemen: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
- CAS-nummer:77-09-8
- Formule:C₂₀H₁₄O₄
- MDL-nummer:MFCD00005913
- Molecular Weight:318,32 g/mol
Specificaties
Specificaties
- Verpakking:Bottle



