Order Entry
Belgium
Orders LinkContactUsLinkComponent
Acetophenone 99%

Acetophenone 99%

Bottle
Catalog # APOSOR1012200500ML
New Product
Supplier:   Apollo Scientific
Acetophenone 99%
Catalog # APOSOR1012200500ML
Supplier:  Apollo Scientific
New Product
CAS Number:  98-86-2
Acetophenone 99%

Bottle

CAS Number:  98-86-2
Price
57,90 €
Log in for availability
1 * 500 mL
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  Methyl phenyl ketone, Phenyl methyl ketone
  • CAS Number:98-86-2
  • Formula:C₈H₈O
  • MDL Number:MFCD00008724
  • Molecular Weight:120,15 g/mol
Specifications

Specifications

  • Verpakking:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.