Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
N-Methylethylamine 97%

N-Methylethylamine 97%

Bottle
Catalogus # APOSOR30626-250MG
Ontvlambaar
Corrosief
Irriterend
New Product
Leverancier:   Apollo Scientific
N-Methylethylamine 97%
Catalogus # APOSOR30626-250MG
Leverancier:  Apollo Scientific
New Product
CAS-nummer:  624-78-2
N-Methylethylamine 97%

Bottle

CAS-nummer:  624-78-2
Price
31,90 €
Log in voor beschikbaarheid
1 * 250 mg
Product Details & Documents
Synoniemen:  N-Ethylmethylamine
  • CAS-nummer:624-78-2
  • Formule:C₃H₉N
  • MDL-nummer:MFCD00009030
  • Molecular Weight:59.11 g/mol
  • Kokend Pt:36…37 °C
  • Dichtheid:0,688 g/cm³ (25 °C)
  • Flits Pt:–34 °C
  • Temperatuur bij opslag:Koelkast
  • EINECS:210-862-1
  • UN:2733
  • ADR:3+8,II
Specificaties

Specificaties

  • Verpakking:
    Bottle

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.