Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Selenium(IV) oxide 99,8%

Selenium(IV) oxide 99,8%

Bottle
Catalogus # APOSIN9857-100G
New Product
Leverancier:   Apollo Scientific
Selenium(IV) oxide 99,8%
Catalogus # APOSIN9857-100G
Leverancier:  Apollo Scientific
New Product
CAS-nummer:  7446-08-4
Selenium(IV) oxide 99,8%

Bottle

CAS-nummer:  7446-08-4
Price
160,00 €
Log in voor beschikbaarheid
1 * 100 g
Product Details & Documents

Usage: Inorganic salt or catalyst

  • CAS-nummer:7446-08-4
  • Formule:H₂O₂Se
  • MDL-nummer:MFCD00003562
  • Molecular Weight:112,97467 g/mol
  • UN:3283
Specificaties

Specificaties

  • Verpakking:
    Bottle

Specificaties