Orders LinkContactUsLinkComponent
(2-Hydroxypropyl)-beta-cyclodextrin 98%
(2-Hydroxypropyl)-beta-cyclodextrin 98%
Bottle
Catalogus # APOSBI4556-2.5KG
Irriterend
New Product
Leverancier:   Apollo Scientific
(2-Hydroxypropyl)-beta-cyclodextrin 98%
Catalogus # APOSBI4556-2.5KG
Leverancier:  Apollo Scientific
New Product
CAS-nummer:  128446-35-5
(2-Hydroxypropyl)-beta-cyclodextrin 98%
Bottle
CAS-nummer:  128446-35-5
Price
261,00 €
Log in voor beschikbaarheid
1 * 2,5 kg
Product Details & Documents
Usage: Biochemical/life science applications
Synoniemen:  Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD
  • CAS-nummer:128446-35-5
  • Formule:C₆₃H₁₁₂O₄₂
  • MDL-nummer:MFCD00069372
  • Molecular Weight:1541,54 g/mol
  • EINECS:420-920-1
Specificaties
Specificaties
  • Verpakking:
    Bottle