Orders LinkContactUsLinkComponent
Methyl red indicator
Methyl red indicator
Bottle
Catalogus # APOSOR95401-2.5KG
New Product
Leverancier:   Apollo Scientific
Methyl red indicator
Catalogus # APOSOR95401-2.5KG
Leverancier:  Apollo Scientific
New Product
CAS-nummer:  493-52-7
Methyl red indicator
Bottle
CAS-nummer:  493-52-7
Price
281,00 €
Log in voor beschikbaarheid
1 * 2,5 kg
Product Details & Documents
Usage: Organic building block/reagent.
  • Non-hazardous
Synoniemen:  2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
  • CAS-nummer:493-52-7
  • Formule:C₁₅H₁₅N₃O₂
  • MDL-nummer:MFCD00002425
  • Molecular Weight:269,3 g/mol
  • EINECS:207-776-1
Specificaties
Specificaties
  • Verpakking:
    Bottle
Specificaties