Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Acetophenone ≥98%

Bottle
  APOSBUP18875-5G
New Product
Acetophenone ≥98%
  APOSBUP18875-5G
New Product
 :  98-86-2
Acetophenone ≥98%

Bottle

 :  98-86-2
 
165,00 €
   
1 * 5 g
 

Usage: Biochemical/ life science applications.

 :  Methyl phenyl ketone, Phenyl methyl ketone
  •  :98-86-2
  •  :C₈H₈O
  •  :120,15 g/mol
 

 

  • Verpakking:
    Bottle