Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

D-(-)-N-Methylglucamine

Bottle
Catalog # APOSOR5855-1KG
Irritant
New Product
Supplier:   Apollo Scientific
D-(-)-N-Methylglucamine
Catalog # APOSOR5855-1KG
Supplier:  Apollo Scientific
New Product
CAS Number:  6284-40-8
D-(-)-N-Methylglucamine

Bottle

CAS Number:  6284-40-8
Price
158,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Meglumine, (-)-1-Deoxy-1-(methylamino)-D-glucitol, N-Methyl-D-glucamine, (-)-N-Methyl-D-glucamine, (2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol, (-)-1-Methylamino-1-deoxy-D-glucitol
  • CAS Number:6284-40-8
  • Formula:H₃CNHCH₂(CH(OH))₄CH₂OH
  • MDL Number:MFCD00004707
  • Molecular Weight:195.22 g/mol
  • Melting Pt:129…131,5 °C
  • Storage Temperature:Kamertemperatuur
  • EINECS:228-506-9
Specifications

Specifications

  • Verpakking:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.