Chlorotoluron
Chlorotoluron
Catalogus # EHERC11530000
Leverancier: EHRENSTORFER
Ga naar:
Product Details & Documents
- SMILES: CN(C)C(=O)Nc1ccc(C)c(Cl)c1
- Inchi: InChI=1S/C10H13ClN2O/c1-7-4-5-8(6-9(7)11)12-10(14)13(2)3/h4-6H,1-3H3,(H,12,14)
- IUPAC: 3-(3-chloro-4-methylphenyl)-1,1-dimethylurea
- Packing group: III
Specificaties
Specificaties
- Omschrijving:Chlorotoluron
- UN No.:3077
- Cat. No.:EHERC11530000
- Moleculair gewicht:212,68
- GHS-signaal:Gevaar
- Beschrijving van de gevarenklasse:GHS08, GHS09
- Formule:C₁₀H₁₃ClN₂O
- Synonym(s):Chlortoluron
- CAS No.:15545-48-9
- Opslag condities:20 °C
- Shipping conditions:RT
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



